
Copper(I) trifluoromethanesulfonate toluene complex

Synonym(s):
Copper(I) triflate toluene complex, Cuprous trifluoromethanesulfonate toluene complex, Trifluoromethanesulfonic acid copper(I) salt toluene complex
Linear Formula:
(CF3SO3Cu)2 · C6H5CH3
CAS Number:
Molecular Weight:
517.37
Products
Sustainability
Feature
Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing