
Clozapine

Synonym(s):
8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]-diazepine
Empirical Formula (Hill Notation):
C18H19ClN4
CAS Number:
Molecular Weight:
326.82
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing