
Chicoric Acid

Synonym(s):
2,3-Dicaffeoyl-L-tartaric acid, Cichoric acid
Empirical Formula (Hill Notation):
C22H18O12
CAS Number:
Molecular Weight:
474.37
Products
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing