
Celecoxib impurity B

Synonym(s):
4-[3-(4-Methylphenyl)-5-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide
Empirical Formula (Hill Notation):
C17H14F3N3O2S
CAS Number:
Molecular Weight:
381.37
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing