
Calcium acetate hydrate

Synonym(s):
Calcium acetate hydrate
Linear Formula:
(CH3COO)2Ca · x H2O
CAS Number:
Molecular Weight:
158.17 (anhydrous basis)
Products
Product Category
Brand
Color
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Showing 1-4 of 4