
Boron trichloride solution

Empirical Formula (Hill Notation):
BCl3
CAS Number:
Molecular Weight:
117.17
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Reaction Type
Reagent Type
Core
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing