Skip to Content
Merck
Bis(pentafluorophenyl) carbonate 97%

Bis(pentafluorophenyl) carbonate

Bis(pentafluorophenyl) carbonate 97%
Synonym(s):
Pentafluorophenyl carbonate
Linear Formula:
(C6F5O)2CO
CAS Number:
Molecular Weight:
394.12

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing