
Bavachinin

Synonym(s):
4′-Hydroxy-7-methoxy-6-(3-methyl-2-butenyl)flavanone, 7-O-Methylbavachin, Bavachinin A
Empirical Formula (Hill Notation):
C21H22O4
CAS Number:
Molecular Weight:
338.40
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing