
BAPTA-AM

Synonym(s):
1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl ester), BAPTA/AM
Empirical Formula (Hill Notation):
C34H40N2O18
CAS Number:
Molecular Weight:
764.68
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing