
Aristolochic acid I

Synonym(s):
TR 1736
Empirical Formula (Hill Notation):
C17H11NO7
CAS Number:
Molecular Weight:
341.27
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3