
Anagrelide Related Compound A

Synonym(s):
Ethyl 2-(6-amino-2,3-dichlorobenzylamino)acetate hydrochloride, Ethyl N-(6-amino-2,3-dichlorobenzyl)glycine hydrochloride
Empirical Formula (Hill Notation):
C11H14Cl2N2O2 · xHCl
CAS Number:
Molecular Weight:
277.15 (free base basis)
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2