
Ammonium molybdate

Synonym(s):
Ammonium molybdate tetrahydrate, Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate
Linear Formula:
(NH4)6Mo7O24 · 4H2O
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Agency
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing