
Aminocarb

Synonym(s):
(4-Dimethylamino-3-methylphenyl) N-methylcarbamate
Empirical Formula (Hill Notation):
C11H16N2O2
CAS Number:
Molecular Weight:
208.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2