
Acenaphthene-d10 solution

Empirical Formula (Hill Notation):
C12D10
CAS Number:
Molecular Weight:
164.27
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing