
Acemetacin

Synonym(s):
1-(p-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic acid carboxymethyl ester
Empirical Formula (Hill Notation):
C21H18ClNO6
CAS Number:
Molecular Weight:
415.82
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing