
Abacavir Related Compound C

Synonym(s):
(1S,4R)-4-(2-Amino-6-chloro-9H-purin-9-yl)-2-cyclopentene-1-methanol hydrochloride
Empirical Formula (Hill Notation):
C11H12ClN5O · HCl
CAS Number:
Molecular Weight:
302.16
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2