
α-Amyl-trans-cinnamaldehyde

Synonym(s):
2-Benzylideneheptanal
Empirical Formula (Hill Notation):
C14H18O
CAS Number:
Molecular Weight:
202.29
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing