Skip to Content
Merck
9-Methyl-9H-fluorene-9-carbonyl chloride ≥99.0% (GC)

9-Methyl-9H-fluorene-9-carbonyl chloride

9-Methyl-9H-fluorene-9-carbonyl chloride ≥99.0% (GC)
Synonym(s):
9-Methylfluorene-9-carbonyl chloride
Empirical Formula (Hill Notation):
C15H11ClO
CAS Number:
Molecular Weight:
242.70

Products

Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing