
6-Carboxyfluorescein

Synonym(s):
6-FAM, FLUOS, 6-Carboxyfluorescein
Empirical Formula (Hill Notation):
C21H12O7
CAS Number:
Molecular Weight:
376.32
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reagent Type
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing