
3,5-Bis(trifluoromethyl)benzylamine

Linear Formula:
(CF3)2C6H3CH2NH2
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing