Skip to Content
Merck
2-Phenoxyethanol analytical standard

2-Phenoxyethanol

2-Phenoxyethanol analytical standard
Synonym(s):
Ethylene glycol monophenyl ether, Phenylglycol
Linear Formula:
C6H5OCH2CH2OH
CAS Number:
Molecular Weight:
138.16
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3