
1-o-Tolyl-1H-pyrazole-4-carbaldehyde

Synonym(s):
1-(2-Methylphenyl)-1H-pyrazole-4-carbaldehyde
Empirical Formula (Hill Notation):
C11H10N2O
Molecular Weight:
186.21
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing