Skip to Content
Merck
1-Hexadecanethiol ≥95.0% (GC)

1-Hexadecanethiol

1-Hexadecanethiol ≥95.0% (GC)
Synonym(s):
Cetyl mercaptan, Hexadecyl mercaptan, Mercaptan C16, 1-Hexadecanethiol
Linear Formula:
CH3(CH2)15SH
CAS Number:
Molecular Weight:
258.51

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing