Skip to Content
Merck
All Photos(1)

Key Documents

G1007615

LC/MS Analysis of Nucleotides on SeQuant ZIC-cHILIC

application for LC-MS

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

LC/MS: suitable

test parameters

column: SeQuant ZIC-cHILIC, 10 cm x 2.1 mm, 3.0 μm particles (1.50657)
mobile phase: [A] acetonitrile; [B] 50 mM ammonium acetate, pH 5.0 with acetic acid in water
gradient: 26 to 27% B in 10 min; to 27 to 35% B in 10 min; to 26% B in 0.1 min; hold at 26% B for 9.9 min
flow rate: 0.3 mL/min
pressure: 1029 psi (71 bar)
column temp.: 50 °C
detector: Q-TOF (ESI-negative), XIC of the following m/z values: AMP-346, ADP-426, ATP-506, CMP-322, CDP-402, (CTP-482, GMP-362, GDP-442, GTP-522, UMP-323, UDP-403, UTP-483)
injection: 10 μL
sample: 10-30 μg/mL of nucleotides (AMP, ADP, ATP, CMP, CDP, CTP, GMP, GDP, GTP, UMP, UDP, UTP) (80/20 acetonitrile/50 mM ammonium acetate in water, pH 5.3 with acetic acid)

suitability

application for LC-MS

application(s)

pharmaceutical (small molecule)

Analysis Note

Nucleotides are the building blocks of nucleic acids. They comprise a nitrogenous base, a sugar, and a phosphate group making them highly polar and ionic. This causes a challenge for typical reversed phase chromatography (RPC). These charged polar analytes are poorly retained by RPC, and thus typically chromatographed by either ion-exchange chromatography or RPC in conjunction with an ion-pairing reagent. Nucleotide analysis using ion-pair reagents or high concentration of phosphate buffers cause high background and ion source pollution for MS detection. HILIC separation is an alternative that permits sensitive MS detection and without the use of ion-pair reagents.

Legal Information

SeQuant is a trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Adenosine 5′-monophosphate sodium salt, from yeast, ≥99%
    Adenosine 5′-diphosphate sodium salt, bacterial, ≥95% (HPLC)
    Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial
    Cytidine 5′-monophosphate, Sigma Grade, ≥99% (HPLC), synthetic, powder
    Cytidine 5′-diphosphate sodium salt hydrate, from yeast, ≥95%
    Cytidine 5′-triphosphate disodium salt, ~95%
    Guanosine 5′-monophosphate disodium salt hydrate, from yeast, ≥99%
    Guanosine 5′-diphosphate sodium salt, Type I, ≥96% (HPLC)
    Guanosine 5′-triphosphate sodium salt hydrate, ≥95% (HPLC), powder
    Uridine 5′-monophosphate, ≥98%
    Uridine 5′-(trihydrogen diphosphate) sodium salt from Saccharomyces cerevisiae, 95-100%
    Uridine 5′-triphosphate trisodium salt hydrate, from yeast, Type III, ≥96%
    See All (12)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service