Skip to Content
Merck
All Photos(1)

Key Documents

98936

Supelco

Methacrylamide

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CONH2
CAS Number:
Molecular Weight:
85.10
Beilstein:
605397
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.21

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

technique(s)

HPLC: suitable

mp

106-110 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CC(=C)C(N)=O

InChI

1S/C4H7NO/c1-3(2)4(5)6/h1H2,2H3,(H2,5,6)

InChI key

FQPSGWSUVKBHSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 - STOT SE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service