Skip to Content
Merck
All Photos(1)

Documents

SRP3079

Sigma-Aldrich

IL-16 human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

LCF (Lymphocyte Chemoattractant Factor)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

12.4 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL16(3603)

General description

IL-16 (interleukin 16), or LCF (lymphocyte chemoattractant factor), was first functionally and biochemically characterized in 1982. IL-16 is produced as a precursor protein (pro-IL16), composed of 631 residues which undergoes cleavage by caspase-3 to produce and secrete a C-terminal biologically active peptide of 121 amino acids. This gene is localized to human chromosome 15q26.3.

Biochem/physiol Actions

IL-16 (interleukin 16) is a pleiotropic proinflammatory cytokine that stimulates chemotactic and immunomodulatory responses of CD4+ (cluster of differentiation) lymphocytes, monocytes, and eosinophils, via signaling through CD4. It prevents HIV (human immunodeficiency virus) replication, and promotes the expression of IL-2R by human T-cells. Study in Chinese population show that polymorphisms in this gene are linked with susceptibility to knee osteoarthritis (OA). IL-16 plays a key role in the release of tumor-related inflammatory cytokines, and the rs4778889 variant in this gene is linked with increased risk of RCC (renal cell cancer) in Chinese population.
Human and murine IL-16 show significant cross-species reactivity. Recombinant human IL-16 is a 12.4kDa protein consisting of 121 amino acid residues.

Sequence

SAASASAASD VSVESTAEAT VCTVTLEKMS AGLGFSLEGG KGSLHGDKPL TINRIFKGAA SEQSETVQPG DEILQLGGTA MQGLTRFEAW NIIKALPDGP VTIVIRRKSL QSKETTAAGD S

Physical form

Lyophilized from 5 mM Sodium Citrate, pH 5.5 + 0.5 mM DTT.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL-16 rs4778889 polymorphism contribution to the development of renal cell cancer in a Chinese population.
Yang SX et al
Genetics and molecular research : GMR, 15(2) (2016)
Conservation of structure and function between human and murine IL-16.
Keane J et al
Journal of Immunology, 160(12), 5945-5954 (1998)
Assignment of human interleukin 16 (IL16) to chromosome 15q26.3 by radiation hybrid mapping.
Kim HS
Cytogenetics and Cell Genetics, 84(1-2), 93-93 (1999)
Center, D.M., et al.
Cytokine Reference, 226-240 (2001)
W W Cruikshank et al.
Journal of immunology (Baltimore, Md. : 1950), 157(12), 5240-5248 (1996-12-15)
We have recently described the cDNA and predicted protein structure of a natural soluble CD4 ligand, IL-16. IL-16 is chemotactic for CD4+ T cells and induces functional IL-2 receptors in CD4+ T cells. The binding of IL-16 to CD4 results

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service