Skip to Content
Merck
All Photos(1)

Documents

SML1708

Sigma-Aldrich

JNJ-47965567

≥98% (HPLC)

Synonym(s):

2-(Phenylthio)-N-[[tetrahydro-4-(4-phenyl-1-piperazinyl)-2H-pyran-4-yl]methyl]-3-pyridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32N4O2S
CAS Number:
Molecular Weight:
488.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NCC1(CCOCC1)N2CCN(C3=CC=CC=C3)CC2)C4=CC=CN=C4SC5=CC=CC=C5

InChI

1S/C28H32N4O2S/c33-26(25-12-7-15-29-27(25)35-24-10-5-2-6-11-24)30-22-28(13-20-34-21-14-28)32-18-16-31(17-19-32)23-8-3-1-4-9-23/h1-12,15H,13-14,16-22H2,(H,30,33)

InChI key

XREFXUCWSYMIOG-UHFFFAOYSA-N

Biochem/physiol Actions

JNJ-47965567 is a potent P2X7 antagonist with high affinity for the rat receptor (pKi = 8.7). It is centrally available after systemic injection with a superior brain:plasma distribution compared to other available P2X7 antagonists. JNJ-47965567 was shown to suppress epileptic seizures in a mouse model of epilepsy. It appears to have a disease modifying effect since spontaneous seizure rates did not increase once treatment with JNJ-477965567 was stopped.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alba Jimenez-Pacheco et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(22), 5920-5932 (2016-06-03)
Neuroinflammation is thought to contribute to the pathogenesis and maintenance of temporal lobe epilepsy, but the underlying cell and molecular mechanisms are not fully understood. The P2X7 receptor is an ionotropic receptor predominantly expressed on the surface of microglia, although
Anindya Bhattacharya et al.
British journal of pharmacology, 170(3), 624-640 (2013-07-31)
An increasing body of evidence suggests that the purinergic receptor P2X, ligand-gated ion channel, 7 (P2X7) in the CNS may play a key role in neuropsychiatry, neurodegeneration and chronic pain. In this study, we characterized JNJ-47965567, a centrally permeable, high-affinity
Juan Mauricio Garré et al.
Nature communications, 11(1), 1784-1784 (2020-04-15)
Dysregulated immunity has been implicated in the pathogenesis of neurodevelopmental disorders but its contribution to synaptic and behavioral deficits in Rett syndrome (RTT) remains unknown. P2X7 receptors (P2X7Rs) are unique purinergic receptors with pro-inflammatory functions. Here, we report in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service