P3819
Phytanic acid methyl ester
≥95%
Synonym(s):
3,7,11,15-Tetramethylhexadecanoic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H42O2
CAS Number:
Molecular Weight:
326.56
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
biological source
synthetic (organic)
Quality Level
Assay
≥95%
form
liquid
functional group
ester (methyl)
lipid type
saturated FAs
shipped in
ambient
storage temp.
2-8°C
SMILES string
COC(=O)CC(C)CCCC(C)CCCC(C)CCCC(C)C
InChI
1S/C21H42O2/c1-17(2)10-7-11-18(3)12-8-13-19(4)14-9-15-20(5)16-21(22)23-6/h17-20H,7-16H2,1-6H3
InChI key
LAWJUFPULQZGLF-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Mohamed M Elmazar et al.
PloS one, 8(1), e45638-e45638 (2013-01-10)
Since activation of PPARγ is the main target for the antidiabetic effect of TZDs, especially when it heterodimerizes with RXR, we aimed to test the potential antidiabetic effect of phytol (250 mg/kg), the natural precursor of phytanic acid, a RXR
Avery L McIntosh et al.
Archives of biochemistry and biophysics, 635, 17-26 (2017-10-21)
While prior studies focusing on male mice suggest a role for sterol carrier protein-2/sterol carrier protein-x (SCP-2/SCP-x; DKO) on hepatic phytol metabolism, its role in females is unresolved. This issue was addressed using female and male wild-type (WT) and DKO
Stephen M Storey et al.
Journal of lipid research, 58(6), 1153-1165 (2017-04-16)
Studies in vitro have suggested that both sterol carrier protein-2/sterol carrier protein-x (
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service