Skip to Content
Merck
All Photos(1)

Key Documents

L4388

Sigma-Aldrich

Calcium L-lactate hydrate

powder, BioReagent, suitable for cell culture

Synonym(s):

(S)-(+)-2-Hydroxypropanoic acid hemicalcium salt, L-Lactic acid hemicalcium salt, Sarcolactic acid hemicalcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3CH(OH)COO]2Ca · xH2O
CAS Number:
Molecular Weight:
218.22 (anhydrous basis)
Beilstein:
4923285
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic

product line

BioReagent

Assay

≥85%

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 50 mg/mL

storage temp.

2-8°C

SMILES string

O.C[C@H](O)C(=O)O[Ca]OC(=O)[C@H](C)O

InChI

1S/2C3H6O3.Ca.H2O/c2*1-2(4)3(5)6;;/h2*2,4H,1H3,(H,5,6);;1H2/q;;+2;/p-2/t2*2-;;/m00../s1

InChI key

MDAVASCOAJMZHZ-NVKWYWNSSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anja Nilsen et al.
Translational oncology, 12(3), 576-584 (2019-01-21)
MicroRNA (miRNA) expressions in tumor biopsies have shown potential as biomarkers in cervical cancer, but suitable reference RNAs for normalization of reverse transcription quantitative polymerase chain reaction (RT-qPCR) assays in patient cohorts with different clinicopathological characteristics are not available. We
Zheng-Wen Nie et al.
Toxicology in vitro : an international journal published in association with BIBRA, 61, 104635-104635 (2019-09-04)
Thiamethoxam (TMX) is a neonicotinoid insecticide, the residues of which have been detected on various crops. In addition to its specific acetylcholine toxicity to insects, TMX was also found to be toxic to mammals. Moreover, oocytes are vulnerable to reactive
Long-Feng Lu et al.
Fish & shellfish immunology, 102, 449-459 (2020-05-15)
Calcium (Ca) is a messenger that regulates a multitude of physiological processes, but its functions in antiviral progress remain undefined. In this study, we found that Ca2+ enhances fish survival to defend against spring viraemia of carp virus (SVCV) infection
Nevena Grdović et al.
PloS one, 9(7), e101172-e101172 (2014-07-06)
The diabetes prevention paradigm envisages the application of strategies that support the maintenance of appropriate β-cell numbers. Herein we show that overexpression of CXC chemokine ligand12 (CXCL12) considerably improves the viability of isolated rat Langerhans islet cells and Rin-5F pancreatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service