Skip to Content
Merck
All Photos(1)

Key Documents

FG0415

Sigma-Aldrich

DMEM - low glucose

With stable glutamine, sodium pyruvate and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME, DMEM Low Glucose, Dulbecco′s Eagle Medium, Dulbecco′s Modified Eagle′s Medium - high glucose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
Pricing and availability is not currently available.

Product Name

Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, stable glutamine, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered
sterile

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

glucose: low
HEPES: no
sodium pyruvate: yes
NaHCO3: yes
stable glutamine: yes
phenol red: yes

shipped in

ambient

storage temp.

2-8°C

General description

This DMEM-Low glucose medium is a 1x complete medium with sodium pyruvate added. It also differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media.

Application

Dulbecco′s Modified Eagle′s Medium-low glucose is a fully supplemented liquid form of the original Dulbecco′s formulation, with one key exception. Pyridoxal, which is an aldehyde, has been replaced with pyridoxine. This modification has been shown to improve the stability of the medium.
Dulbecco′s Modified Eagle′s Medium - low glucose has been used in the maintenance/culturing of mammalian cells.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service