Skip to Content
Merck
All Photos(2)

Key Documents

CLS354277

Corning® Matrigel® hESC-Qualified Matrix

LDEV-Free, pack of 5 mL

Synonym(s):

Matrigel Matrix, hESC-Qualified Matrigel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116155
NACRES:
NA.7

feature

LDEV-Free

packaging

pack of 5 mL

manufacturer/tradename

Corning 354277

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Quality

Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
Extensive PCR testing is performed to screen for a number of pathogens
Tested and found negative for bacteria, fungi, and mycoplasma
Protein concentrations determined by Lowry method
Endotoxin units measured by Limulus Amoebocyte Lysate assay
Matrigel Matrix gel stability is tested for a period of 14 days at 37°C
Biological activity is determined for each lot using a neurite outgrowth assay.
Source
Engelbreth-Holm-Swarm (EHS) mouse tumor

Preparation and Storage
Store at -20°C. Avoid multiple freeze-thaws.

Click here for Corning Matrigel Lot Selection and Reservation.

For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.

Legal Information

Corning is a registered trademark of Corning, Inc.
Matrigel is a registered trademark of Corning, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogela are the most widely used systems for 3D cell culture. Learn more about this technology (what are hydrogels? How to chose?)

Protocols

Learn how the Corning® Matrigel® Basement Membrane Matrix can be used to boost your 3D Cell Culture activities.

Learn the results that come from changing the protein concentration of Corning® Matrigel®.

Learn how the Corning® Matrigel® Basement Membrane Matrix can be used to boost your 3D Cell Culture activities.

Learn how the Corning® Matrigel® Basement Membrane Matrix can be used to boost your 3D Cell Culture activities.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service