Skip to Content
Merck
All Photos(1)

Key Documents

IPI-1

93100621, Porcine intestine, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

IPI-1, 93100621

biological source

Porcine intestine

growth mode

Adherent

karyotype

Heteroploid

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Boar Miniature male ileum, SV40 transformed

Cell Line Description

Fibroblast cell line derived from the ileum of an adult histocompatible miniature male boar. Transformed by SV40 plasmid pSV3-neo.

Application

Potential feeder cell line for the recovery of epithelial cells isolated from the small intestine.

Culture Medium

DMEM + 4mM Glutamine + 0.024IU/ml Insulin + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service