Skip to Content
Merck
All Photos(1)

Key Documents

C-39635

Sigma-Aldrich

Chondrocyte Growth Medium

SupplementMix containing all media supplements pre-mixed in one vial, 1 Mix for 500 ml

Synonym(s):

Recommended with C-27111

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

aqueous solution

packaging

pkg of 1 vial

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Chondrocyte Growth Medium SupplementMix (basal medium sold separately)

To form a complete medium, it should be combined with the basal medium: C-27111

Application

Chondrocyte Growth Medium has been developed for the in vitro cultivation of human chondrocytes. The fetal calf serum in the medium is strictly pretested for optimal performance. The medium is optimized for primary human cells, but can also be used for primary porcine and rat chondrocytes.Recommended for:Human Chondrocytes (HCH)

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Chondrocyte Growth Medium are subjected to comprehensive quality control tests using primary human chondrocytes. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested chondrocytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Storage and Stability

Store the SupplementMix at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

recommended

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service