Skip to Content
Merck
All Photos(1)

Key Documents

LTN-AHRE-1

LoTox Peanut Flour Protein with Defined Allergen Content (Arachis hypogaea), peanut

pkg of 1 mL, storage temp.:-20°C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

packaging

pkg of 1 mL

manufacturer/tradename

InBio

technique(s)

HPLC: suitable
LC/MS: suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

General description

Produced and qualified by Indoor Biotechnologies.

Source: Light roasted peanut flour (Runner cultivar)
Formulation: Preservative and carrier-free in sterile, endotoxin-free Phosphate-buffered saline, pH 7.4. Sterile filtered.
Concentration: See product insert.
Purification:

Note:
(1) A LoTox­ product: Endotoxin = 0.03 EU/μg.
(2) Use sterile tips to avoid contamination of product.
(3) Avoid repeated Freeze/Thaw cycles. It is recommended to aliquot the product upon first thaw and freeze in working aliquot volumes.

Storage and Stability

Do not allow product to remain at 4°C or room temperature for extended periods of time. Re-freeze unused portion and aliquot into smaller quantities if necessary to avoid freeze/thaw cycles.

Analysis Note

Ara h 1: 64 kD monomer; Ara h 2: 19-19 kD doublet
Ara h 3: 58 kD non-red; Ara h 6: 14.5 kD; Ara h 8: 17 kD

Disclaimer

Allergens are provided for research and commercial use in vitro:  not for human in vivo or therapeutic use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service