Skip to Content
Merck
All Photos(4)

Documents

64340

Sigma-Aldrich

DL-Methionine

≥99.0% (NT)

Synonym(s):

(±)-2-Amino-4-(methylmercapto)butyric acid, DL-2-Amino-4-(methylthio)butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
149.21
Beilstein:
636185
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

DL-Methionine, ≥99.0% (NT)

Quality Level

Assay

≥99.0% (NT)

form

powder or crystals

optical activity

[α]/D, c = 5 in 5 M HCl (inactive)

concentration

5% in 5 M HCl

ign. residue

≤0.05%

color

white

mp

280 °C (dec.) (lit.)

solubility

5 M HCl: 1 g/10 mL, clear, colorless to almost colorless

suitability

corresponds for TLC

application(s)

peptide synthesis

SMILES string

CSCCC(N)C(O)=O

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)

InChI key

FFEARJCKVFRZRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DL-Methionine has been used to study the susceptibility of methionine to oxidation during the development of monoclonal antibodies.

Biochem/physiol Actions

Methionine is an essential amino acid which acts a precursor for transmethylation and transsulfuration. In proteins, L-methionine is the biologically active form. D-methionine is converted to L-methionine by enzymes in the body. The adenosylation of methionine results in the formation of S-adenosyl-L-methionine (SAM), which serves as a methyl donor to various substances. L-Methionine′s metabolic product glutathione is known to regulate immune response and has antiviral action.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid assessment of oxidation via middle-down LCMS correlates with methionine side-chain solvent-accessible surface area for 121 clinical stage monoclonal antibodies
Rong Yang
MAbs (2017)
L-methionine as immune supportive supplement: a clinical evaluation.
Van Brummelen and du Toit D
Amino Acids, 33(1), 157-163 (2007)
Il Jin Cho et al.
Biochemical and biophysical research communications, 367(1), 190-194 (2008-01-02)
We investigated the hypolipidemic effect of resveratrol focused on the mRNA expression and hepatic HMG-CoA reductase (HMGR) activity in hamsters fed a high-fat diet. Male Syrian Golden hamsters were fed a high-fat diet containing 0.025% fenofibrate or 0.025% resveratrol for
Bio-Methionine Production from Glucose - Cost Analysis - Methionine E31A (2016)
Pilar Lloris-Garcerá et al.
Journal of molecular biology, 426(11), 2246-2254 (2014-04-03)
The increasing number of solved membrane protein structures has led to the recognition of a common feature in a large fraction of the small-molecule transporters: inverted repeat structures, formed by two fused homologous membrane domains with opposite orientation in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service