Skip to Content
Merck
All Photos(1)

Key Documents

61407

Supelco

Trigonelline hydrochloride

analytical standard

Synonym(s):

1-Methylpyridinium-3-carboxylate hydrochloride, N-Methylnicotinic acid betaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2 · HCl
CAS Number:
Molecular Weight:
173.60
Beilstein:
3734169
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

food and beverages

format

neat

SMILES string

[Cl-].C[n+]1cccc(c1)C(O)=O

InChI

1S/C7H7NO2.ClH/c1-8-4-2-3-6(5-8)7(9)10;/h2-5H,1H3;1H

InChI key

TZSYLWAXZMNUJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanna Makowska et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 968, 101-104 (2014-01-01)
For trigonelline, a quaternary-base pyridine alkaloid of presumed Alzheimer's disease-preventing activity, a method of determination has been proposed, based on the hydrophilic interaction chromatography (HILIC). That method might be applied to study the agent's bioavailability, in particular its permeation through
Kouichi Mizuno et al.
Biochemical and biophysical research communications, 452(4), 1060-1066 (2014-09-23)
Trigonelline (N-methylnicotinate), a member of the pyridine alkaloids, accumulates in coffee beans along with caffeine. The biosynthetic pathway of trigonelline is not fully elucidated. While it is quite likely that the production of trigonelline from nicotinate is catalyzed by N-methyltransferase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service