Skip to Content
Merck
All Photos(1)

Key Documents

01709

Supelco

Acrylamide:N,N′-Methylenebisacrylamide 37.5:1 solution

suitable for electrophoresis, mixture of acrylamide and BIS 37.5:1, 40% in H2O

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

concentration

40% in H2O

technique(s)

electrophoresis: suitable

impurities

≤0.01% free acrylic acid

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤10 mg/kg
Mg: ≤5 mg/kg
Mn: ≤1 mg/kg
Na: ≤10 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

suitability

in accordance for electrophoresis test

storage temp.

2-8°C

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily F Willis et al.
STAR protocols, 1(3), 100211-100211 (2020-12-31)
The advent of tools enabling the direct manipulation of microglia has furthered our understanding of their role in health and disease. Here, we present a detailed protocol allowing for microglia turnover in adult CX3CR1 creERT2 × iDTR or CX3CR1 creERT2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service