Skip to Content
Merck
All Photos(1)

Key Documents

SCR108

Sigma-Aldrich

Human Mesenchymal Stem Cell Kit - Derived from Bone-Marrow

This Human Mesenchymal Stem Cell Kit contains 1 million viable cells & 500ml of optimized media.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509
eCl@ss:
32161000
NACRES:
NA.81

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable

input

sample type mesenchymal stem cell(s)

shipped in

liquid nitrogen

General description

Product Source: Human bone marrow

Application

Mesenchymal stem cells, also known as marrow stromal cells (1), are defined as a self-renewing population of adherent, bone-marrow-derived multipotent progenitor cells with the capacity to differentiate into several mesenchymal cell lineages. In defined in vitro assays, mesenchymal stem cells have been shown to readily differentiate into lineage-specific cells that form bone, cartilage, fat, tendon, and muscle tissues (1, 2). Mesenchymal stem cells also provide support and maintenance for the other major stem cell population in the bone marrow, the hematopoietic stem cells (2).
Mesenchymal stem cells (MSC’s) have historically been isolated based on the ability of these cells to form adherent cell layers in culture and the concomitant lack of adherence of other cells in the bone marrow stroma such as hematopoietic stem cells, adipocytes, and macrophage (1, 3). While this procedure results in enriched populations of mesenchymal stem cells, the resulting bone-marrow derived cell populations are, nonetheless, heterogeneous – comprised not only of mesenchymal stem cells, but also of committed lineage-restricted progenitors (1, 3). To aid researchers in the accurate identification and characterization of mesenchymal stem cells, Millipore provides cryopreserved Human Mesenchymal Stem Cells and optimized media in this kit. Millipore’s cryopreserved Human Mesenchymal Stem Cells are ready-to-use primary mesenchymal stem cells isolated from the iliac crest of normal human bone marrow (18-30 years old).
Research Category
Stem Cell Research
This Human Mesenchymal Stem Cell Kit contains 1 million viable cells & 500ml of optimized media.

Quality

Millipore’s Human Mesenchymal Stem Cells are derived from human bone marrow and have been validated for high expression level of cell surface molecules that are present on mesenchymal stem cells: H-CAM (CD44), THY-1 (CD90) and STRO-1 and for their absence of hematopoietic cell surface markers, CD14 (present on leukocytes) and CD19 (present on B-lymphocytes). Millipore’s Human Mesenchymal Stem Cells are also negative for the endothelial marker, M-CAM (CD146). The cells have also been validated for their self-renewal and multi-lineage differentiation capacities (please refer to product manual figures for representative data). Cells display normal karyotype as assessed by G-banding of 20 metaphase cells and tested negative for mycoplasma. Each lot of Mesenchymal Stem Cell medium is tested for the optimal cell growth and proliferation of mesenchymal stem cells. Sterility Testing: Negative

Storage and Stability

Human Mesenchymal Stem Cells (Part # SCC034) should be stored in liquid nitrogen.
Mesenchymal Stem Cell Expansion Medium: (Catalog No. SCM015) should be stored at -20°C until ready to use. Upon thawing, the basal medium should be stored at 2-8°C for up to one month.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert G Gourdie et al.
Microscopy and microanalysis : the official journal of Microscopy Society of America, Microbeam Analysis Society, Microscopical Society of Canada, 18(1), 99-106 (2012-01-05)
A novel self-organizing behavior of cellularized gels composed of collagen type 1 that may have utility for tissue engineering is described. Depending on the starting geometry of the tissue culture well, toroidal rings of cells or hollow spheroids were prompted
Angel T Chan et al.
Biomaterials, 73, 1-11 (2015-09-18)
Cell death due to anoikis, necrosis and cell egress from transplantation sites limits functional benefits of cellular cardiomyoplasty. Cell dissociation and suspension, which are a pre-requisite for most cell transplantation studies, lead to depression of cellular metabolism and anoikis, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service