Skip to Content
Merck
All Photos(1)

Key Documents

MABS1966

Sigma-Aldrich

Anti-PEG Antibody, clone 6.3

clone 6.3, from mouse

Synonym(s):

Polyethylene Glycol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6.3, monoclonal

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable

isotype

IgG1κ

target post-translational modification

unmodified

General description

Polyethylene glycols (PEG) are non-toxic, uncharged, hydrophilic long chain polymers with a big exclusion volume in aqueous solution. The covalent attachment of PEG is commonly employed to modify proteins and drugs that can increase their bioavailability and reduce immunogenicity and proteolytic degradation. Clone 6.3 displays high affinity and binds the PEG backbone and can be employed as a detection antibody for the quantification of PEG. It binds to a smaller epitope of the PEG backbone when compared to other antibodies. Since it binds to a smaller PEG epitope (~750 Da), it is not compatible with detergents that contain an ethylene oxide backbone, such as Tween 20. Instead 0.05% CHAPS is a more suitable detergent when using this antibody.

Specificity

Clone 6.3 specifically detects proteins conjugated with polyethylene glycol (PEG).

Immunogen

PEG-conjugated to E.coli beta-glucuronidase.

Application

Anti-PEG, clone 6.3, Cat. No. MABS1966, is a mouse monoclonal antibody that detects PEG-conjugated proteins and it hs been tested for use in ELISA.
ELISA Analysis: A representative lot detected PEG in ELISA applications (Tung, H.Y., et. al. (2015). Mol Cancer Ther. 14(6):1317-26).
Research Category
Signaling

Quality

Evaluated by ELISA in Immobilized PEG (10K-PEG, 3K-PEG, and 2K-mPEG).

ELISA Analysis: 5 µg/mL of Immobilized PEG (10K-PEG, 3K-PEG, and 2K-mPEG) were detected with this antibody.

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal antibody IgG1 in PBS with 0.04% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service