Skip to Content
Merck
All Photos(3)

Documents

ECM675

Sigma-Aldrich

3D Collagen Culture Kit

Synonym(s):

3D Cell Culture Kit, Collagen Culture Kit, Kit for 3D Collagen Culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32161000
NACRES:
NA.75

species reactivity

human

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

activity assay: suitable
cell based assay: suitable
cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Cell migration is a fundamental function of normal cellular processes, including cell proliferation, and migration. Cell behavior in vitro is typically examined in a 2D environment; however, in the body, cells exist in a three-dimensional extracellular matrix environment rich in type I collagen. In fact, cells cultured in 3D matrices better reflect in vivo cell physiology when compared to traditional 2D systems.

CHEMICON (EMD Millipore) has developed a three-dimensional cell culture system that closely simulates the collagen-rich extracellular matrix of normal tissue. This 3D system provides a simple and rapid method to analyze cell angiogenesis, migration, apoptosis, proliferation and tissue formation in a 3D-collagen matrix. Cells suspended in this 3D system are easily visualized by phase contrast or fluorescence light microscopy. Cells can be directly fixed and stained within the matrix and treated with antibodies for visualization of specific intra- and extracellular proteins. Importantly, cells suspended in our 3D system can also be treated with various reagents allowing for extensive screening of biological responses to growth factors and chemical agents.

As an additional benefit to investigators, sterile and viable cells may be removed from the 3D Cell Culture System for further experimentation, including FACS or biochemical analysis.

For Research Use Only; Not for use in diagnostic procedures

Application

Research Category
Cell Structure
This 3D system provides a simple and rapid method to analyze cell angiogenesis, migration, apoptosis, proliferation and tissue formation in a 3D-collagen matrix. Cells suspended in this 3D system are easily visualized by phase contrast or fluorescence light microscopy. Cells can be directly fixed and stained within the matrix and treated with antibodies for visualization of specific intra- and extracellular proteins. Importantly, cells suspended in our 3D system can also be treated with various reagents allowing for extensive screening of biological responses to growth factors and chemical agents.

As an additional benefit to investigators, sterile and viable cells may be removed from the 3D Cell Culture System for further experimentation, including FACS or biochemical analysis.

Components

Collagen Solution - (Part No. 90135) One bottle - 10 mL

5X RPMI Medium- (Part No. 90134) One bottle - 2.5 mL

5X M199 Medium- (Part No. 90133) One bottle - 2.5 mL

5X DMEM Medium- (Part No. 90137) One bottle - 2.5 mL

5X PBS with Phenol Red- (Part No. 90136) One bottle - 2.5 mL

Neutralization Solution- (Part No. 90138) One vial - 0.5 mL

Physical form

Collagen solution is supplied in 6mM acetate buffer at ~ pH7.0.

Storage and Stability

Store kit materials at 4°C up to their expiration date.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David G Leach et al.
Biomaterials, 163, 67-75 (2018-02-18)
Recent advancements in the field of immunotherapy have yielded encouraging results for the treatment of advanced cancers. Cyclic dinucleotides (CDNs) are a powerful new class of immunotherapy drugs known as STING (Stimulator of Interferon Genes) agonists, currently in clinical trials.
Xiaoji Wang et al.
Biochemical and biophysical research communications, 403(3-4), 264-269 (2010-11-16)
Medulloblastoma (MB) is the most devastating and common pediatric brain tumor. Tumor cells invading into surrounding tissue and disseminating through cerebrospinal fluid make treatment extremely difficult. Identifying the mechanisms of MB cells is therefore imperative for the development of novel
LiMei Wang et al.
PLoS biology, 6(11), e289-e289 (2008-12-11)
The multifunctional signaling protein p75 neurotrophin receptor (p75(NTR)) is a central regulator and major contributor to the highly invasive nature of malignant gliomas. Here, we show that neurotrophin-dependent regulated intramembrane proteolysis (RIP) of p75(NTR) is required for p75(NTR)-mediated glioma invasion
Zhiyong Chen et al.
Brain research, 1368, 71-81 (2010-10-19)
One of the histopathological consequences of a penetrating ballistic brain injury is the formation of a permanent cavity. In a previous study using the penetrating ballistic-like brain injury (PBBI) model, engrafted human amnion-derived multipotent progenitor (AMP) cells failed to survive
Nicoletta Bivi et al.
Bioorganic & medicinal chemistry, 24(4), 759-767 (2016-01-23)
Raloxifene, a selective estrogen receptor modulator (SERM), reduces fracture risk at least in part by improving the mechanical properties of bone in a cell- and estrogen receptor-independent manner. In this study, we determined that raloxifene directly interacts with the bone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service