Skip to Content
Merck
All Photos(1)

Key Documents

ABS310

Sigma-Aldrich

Anti-hCE-1 Antibody

serum, from rabbit

Synonym(s):

Carboxylesterase 1, Liver carboxylesterase 1, Acyl-coenzyme A:cholesterol acyltransferase, ACAT, Brain carboxylesterase hBr1, CE-1, hCE-1, Cocaine carboxylesterase, Egasyn, HMSE, Methylumbelliferyl-acetate deacetylase 1, Monocyte/macrophage serine estera

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CES1(1066)

General description

Carboxylesterase 1 (hCE-1) is also known as Liver carboxylesterase 1, Acyl-coenzyme A:cholesterol acyltransferase, (ACAT), Cocaine carboxylesterase, Methylumbelliferyl-acetate deacetylase 1, Monocyte/macrophage serine esterase, Retinyl ester hydrolase, (REH), Serine esterase 1, and Triacylglycerol hydrolase (TGH). hCE-1 is involved in: the detoxification of xenobiotics and the activation of ester and amide prodrugs. hCE-1 functions to hydrolize the methyl ester group of cocaine to form benzoylecgonine, catalyze the transesterification of cocaine to form cocaethylene and the ethyl esterification of oleic acid to ethyloleate, and is activated by CHAPS. hCE-1 is expressed mostly in liver with lower levels found in the heart and lung.

Specificity

This antibody does not cross react with human CE-2 (Prof. P. Potter, St. Jude Children′s Research Hospital, Memphis).

Immunogen

Linear peptide corresponding to human hCE-1.

Application

Detect Carboxylesterase 1 using this rabbit polyclonal antibody, Anti-hCE-1 Antibody validated for use in western blotting.
Research Category
Signaling
Research Sub Category
Developmental Signaling
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected hCE-1 in 10 µg of human liver tissue lysate.
Western Blotting Analysis: A representative lot from an independent laboratory detected hCE-1 in hCE-1 transfected U373MG cell lysates, and demonstrated a loss of signal in non-transfected and hCE-2 transfected U373MG cell lysates (Prof. P. Potter, St. Jude Children′s Research Hospital, Memphis.).

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected hCE-1 in 10 µg of HepG2 cell lysate.

Target description

~58 kDa observed. A homodimeric trimer of three hCE-1 molecules may be observed at ~190 kDa in some cell lysates (Prof. P. Potter, St. Jude Children′s Research Hospital, Memphis.).

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HepG2 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service