Skip to Content
Merck
All Photos(2)

Key Documents

1.00272

Supelco

2-Propanol

standard for the analysis of tobacco

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.24

vapor density

2.1 (vs air)

Quality Level

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

≥99.0% (n-heptadecane, GC)
≥99.2% ethanol basis (m/m)
≥99.8% (2-propanol)

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

refractive index

n20/D 1.377 (lit.)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

transition temp

flash point 12 °C

density

0.785 g/mL at 25 °C (lit.)

storage temp.

2-25°C

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • 2-Propanol in solvent applications: 2-propanol is used as a solvent in swelling and delamination of inorganic homoionic montmorillonite clay. The study emphasized its utility in water-polar organic mixed solvents, showcasing its importance in industrial processes requiring high-purity solvents (Nakato et al., 2024).

  • 2-Propanol in chromatography: 2-Propanol is used in a static headspace gas chromatography method for quantitating residual solvents in nanoformulations. This reflects its critical role in analytical techniques like chromatography where reagent grade solvents are essential for accurate and reliable results (Kattel and Clogston, 2024).

  • 2-Propanol in biochemistry research: used as a solvent in the amination of heterocycles under acidic conditions. This study points to its utility as an IPA solvent in biochemistry research, enabling innovative approaches to pharmaceutical compound development (Yasuda et al., 2024).


Analysis Note

Assay (2-Propanol) (GC): ≥ 99.8 %
Assay (Ethanol) (m/m): ≥ 99.2 %
Assay (n-Heptadecane) (GC): ≥ 99.0 %
Ethanol (GC/ +/-1%): 2
n-Heptadecane (GC/ +/-1%): 0.3
Water: ≤ 0.1 %

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service