WIMWG10004
Wilmad® NMR tubes 5 mm diam., high throughput
L 103.5 mm, cap: no
Synonym(s):
high throughput NMR tube
About This Item
Recommended Products
material
borosilicate glass tube
feature
cap: no
neck thread push fit
packaging
pack of 100 ea
manufacturer/tradename
Wilmad WG-1000-4
parameter
600 MHz frequency
L
103.5 mm
tube diam.
5 mm
wall thickness
0.43 mm
Related Categories
General description
High Throughput NMR tubes have an averaged camber of 60 micron to guarantee the spectrum resolution for small molecules (MW<250) samples up to 600 MHz.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service