Skip to Content
Merck
All Photos(1)

Key Documents

W6497

D-Glucose-6,6-d2

API for Clinical Studies, 98 atom % D, 98% (CP)

Synonym(s):

Dextrose-6,6-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D2H10O6
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352116

grade

cGMP

isotopic purity

98 atom % D

Assay

98% (CP)

mp

150-152 °C (lit.)

mass shift

M+2

storage temp.

15-25°C

SMILES string

[2H]C([2H])(O)[C@H]1OC(O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i1D2

InChI key

WQZGKKKJIJFFOK-VARZTKRMSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service