Skip to Content
Merck
All Photos(1)

Key Documents

W466620

Sigma-Aldrich

(-)-α-Bisabolol

natural, ≥95%, FG

Synonym(s):

(-)-6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol, Levomenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
FEMA Number:
4666
MDL number:
UNSPSC Code:
12352212
Flavis number:
02.129
NACRES:
NA.05

grade

FG
Kosher
natural

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; woody; spicy; sweet

storage temp.

2-8°C

InChI

1S/C15H26O/c1-12(2)6-5-11-15(4,16)14-9-7-13(3)8-10-14/h6-7,14,16H,5,8-11H2,1-4H3/t14-,15+/m1/s1

InChI key

RGZSQWQPBWRIAQ-CABCVRRESA-N

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service