Skip to Content
Merck
All Photos(1)

Key Documents

MT51350076

Mettler-Toledo electrolyte for reference electrodes

Friscolyt-B, volume 250 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41101900
NACRES:
NB.79

form

liquid

manufacturer/tradename

Mettler-Toledo 51350076

volume

250 mL

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service