Skip to Content
Merck
All Photos(3)

Key Documents

D206539

Sigma-Aldrich

Diphenyl carbonate

ReagentPlus®, 99%

Synonym(s):

Bisphenyl carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)2CO
CAS Number:
Molecular Weight:
214.22
Beilstein:
1074863
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

form

crystals

bp

301-302 °C (lit.)

mp

79-82 °C (lit.)

SMILES string

O=C(Oc1ccccc1)Oc2ccccc2

InChI

1S/C13H10O3/c14-13(15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H

InChI key

ROORDVPLFPIABK-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

334.4 °F - closed cup

Flash Point(C)

168 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service