Skip to Content
Merck
All Photos(1)

Key Documents

755184

Sigma-Aldrich

Indoxyl-4,5,6,7-d4 sulfate potassium salt

≥97 atom % D, ≥97% (CP)

Synonym(s):

Deuterated indoxyl sulfate potassium salt, Potassium 3-indoxyl-4,5,6,7-d4 sulfate, Urinary indican-4,5,6,7-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D4H2KNO4S
CAS Number:
Molecular Weight:
255.33
UNSPSC Code:
12352106
PubChem Substance ID:

isotopic purity

≥97 atom % D

Assay

≥97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

165 °C (lit.)

mass shift

M+4

storage temp.

−20°C

SMILES string

O=S(OC1=CNC2=C([2H])C([2H])=C([2H])C([2H])=C21)([O-])=O.[K+]

InChI

1S/C8H7NO4S.K/c10-14(11,12)13-8-5-9-7-4-2-1-3-6(7)8;/h1-5,9H,(H,10,11,12);/q;+1/p-1/i1D,2D,3D,4D;

InChI key

MDAWATNFDJIBBD-FOMJDCLLSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service