Skip to Content
Merck
All Photos(2)

Documents

438278

Sigma-Aldrich

Triethylmethylammonium chloride

97%

Synonym(s):

MTEACl, Methyltriethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N(Cl)CH3
CAS Number:
Molecular Weight:
151.68
Beilstein:
3912434
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

282-284 °C (lit.)

SMILES string

[Cl-].CC[N+](C)(CC)CC

InChI

1S/C7H18N.ClH/c1-5-8(4,6-2)7-3;/h5-7H2,1-4H3;1H/q+1;/p-1

InChI key

NIUZJTWSUGSWJI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Triethylmethylammonium chloride can be used as an organic template to synthesize zeolites, applicable as catalysts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Neef et al.
Biochemical pharmacology, 33(24), 3991-4002 (1984-12-15)
The influence of an anorganic anion iodide (I-) and an organic anion tetraphenylborate (TPB-) on the hepatic uptake and biliary excretion of three organic cations, triethylmethyl ammonium (TEMA), tripropylmethyl ammonium (TPMA) and tri-n-butylmethyl ammonium (TBuMA) was studied. The compounds were
Size-Controlled Synthesis of Metallosilicates with MTW Structure and Catalytic Performance for Methanol-to-Propylene Reaction.
Taniguchi T, et al.
Catalysis Letters, 146(3), 666-676 (2016)
C Neef et al.
Biochemical pharmacology, 33(24), 3977-3990 (1984-12-15)
Three quaternary ammonium compounds (QACs) with different lipophilicity, triethylmethyl ammonium iodide (TEMA), tripropylmethyl ammonium iodide (TPMA) and tri-n-butylmethyl ammonium iodide (TBuMA) were given as a bolus injection of 10 mumole and 1 mumole in an isolated perfused liver. TPMA and
Effects of crystal morphology, Si/Al ratio and thickness of an MTW zeolite membrane on water/2-propanol separation by pervaporation.
Zhang Y, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 222, 178-184 (2016)
Development of HZSM-12 zeolite for catalytic degradation of high-density polyethylene.
Silva A O, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 244, 1-6 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service