Skip to Content
Merck
All Photos(1)

Key Documents

376531

Sigma-Aldrich

Iodoethane-13C2

99 atom % 13C, contains copper as stabilizer

Synonym(s):

Ethyl-13C2 iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH313CH2I
CAS Number:
Molecular Weight:
157.95
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

contains

copper as stabilizer

refractive index

n20/D 1.512 (lit.)

bp

69-73 °C (lit.)

mp

-108 °C (lit.)

density

1.975 g/mL at 25 °C

mass shift

M+2

storage temp.

2-8°C

SMILES string

[13CH3][13CH2]I

InChI

1S/C2H5I/c1-2-3/h2H2,1H3/i1+1,2+1

InChI key

HVTICUPFWKNHNG-ZDOIIHCHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Muta. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service